Thermo Scientific Chemicals

Cobalt(II) chloride, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 013666.03
1 g, Each
Thermo Scientific Chemicals

Cobalt(II) chloride, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 013666.03
1 g, Each
Quantity
Catalog number: 013666.03
also known as 013666-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7646-79-9
IUPAC Name
λ²-cobalt(2+) dichloride
Molecular Formula
Cl2Co
InChI Key
GVPFVAHMJGGAJG-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Co++]
Appearance (Color)
Blue
Form
Beads
Residual water
< 100 ppm
Total Metal Impurities
0.1% max.
Particle Size
-10 Mesh

Description

Cobalt(II) chloride in vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text