Terbium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Terbium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Terbium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 013672.03
also known as 013672-03
Price (USD)/ Each
88.30
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
88.30
Add to cart
Terbium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Catalog number013672.03
Price (USD)/ Each
88.30
-
Add to cart
Chemical Identifiers
CAS13813-40-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormFlake
Residual water< 100 ppm
Total Rare Earth Oxide Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only