Thermo Scientific Chemicals

Scandium(III) iodide, ultra dry, 99.999% (REO), Thermo Scientific Chemicals

Catalog number: 013687.06
5 g, Each
Thermo Scientific Chemicals

Scandium(III) iodide, ultra dry, 99.999% (REO), Thermo Scientific Chemicals

Catalog number: 013687.06
5 g, Each
Quantity
Catalog number: 013687.06
also known as 013687-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14474-33-0
IUPAC Name
scandium(3+) triiodide
Molecular Formula
I3Sc
InChI Key
HUIHCQPFSRNMNM-UHFFFAOYSA-K
SMILES
[Sc+3].[I-].[I-].[I-]
Form
Powder
Appearance (Color)
White
Residual water
< 100 ppm
Total Rare Earth Oxide Impurities
0.001% max.

Description

Scandium iodide is used in high-intensity lighting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text