Thermo Scientific Chemicals

Chloropentaamminecobalt(III) chloride, Thermo Scientific Chemicals

Catalog number: 013765.04
2 g, Each
Thermo Scientific Chemicals

Chloropentaamminecobalt(III) chloride, Thermo Scientific Chemicals

Catalog number: 013765.04
2 g, Each
Quantity
Catalog number: 013765.04
also known as 013765-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13859-51-3
Assay (unspecified)
Co 23.0% min

Description

Chloropentaamminecobalt(III) chloride is doped with polyvinyl acetate to prepare polymeric films, which enhances the optical conductivity. It is also used to prepare the corresponding hydrogen sulfate complex by reacting with sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text