Chloropentaamminecobalt(III) chloride, Thermo Scientific Chemicals
Chloropentaamminecobalt(III) chloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloropentaamminecobalt(III) chloride, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2 g
10 g
Catalog number 013765.18
also known as 013765-18
Price (USD)/ Each
485.65
Online exclusive
539.00 
Save 53.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
485.65
Online exclusive
539.00 
Save 53.35 (10%)
Add to cart
Chloropentaamminecobalt(III) chloride, Thermo Scientific Chemicals
Catalog number013765.18
Price (USD)/ Each
485.65
Online exclusive
539.00 
Save 53.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13859-51-3
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Co 23.0% min
Chloropentaamminecobalt(III) chloride is doped with polyvinyl acetate to prepare polymeric films, which enhances the optical conductivity. It is also used to prepare the corresponding hydrogen sulfate complex by reacting with sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only