Vanadium foil, 0.127mm (0.005in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Vanadium foil, 0.127mm (0.005in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium foil, 0.127mm (0.005in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

Form: approx. 3.87g/50x100mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 200 mm
50 x 100 mm
Catalog number 013783.GK
also known as 013783-GK
Price (USD)/ Each
375.65
Online exclusive
417.00 
Save 41.35 (10%)
-
Add to cart
Quantity:
100 x 200 mm
Request bulk or custom format
Price (USD)/ Each
375.65
Online exclusive
417.00 
Save 41.35 (10%)
Add to cart
Vanadium foil, 0.127mm (0.005in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number013783.GK
Price (USD)/ Each
375.65
Online exclusive
417.00 
Save 41.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-62-2
IUPAC Namevanadium
Molecular FormulaV
InChI KeyLEONUFNNVUYDNQ-UHFFFAOYSA-N
SMILES[V]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.8% (metals basis)
Appearance (Color)White silver
FormFoil, 0.127mm (0.005in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only