Chromium powder, -200 mesh, 99.94% (metals basis), Thermo Scientific Chemicals
Chromium powder, -200 mesh, 99.94% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium powder, -200 mesh, 99.94% (metals basis), Thermo Scientific Chemicals

Chromium, Cr, CAS Number-7440-47-3, chrome, chromium, elemental, ccris 159, chrome french, compounds, chrom, chrom german, cromo, metal, chromium, metal, 500g, Gray powder, 99.95% (Metals basis), CHEBI:28073, DOT Class: 4.1, Packaging Group: III, 231-157-5, H228
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 013796.36
also known as 013796-36
Price (USD)/ Each
989.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
989.00
Add to cart
Chromium powder, -200 mesh, 99.94% (metals basis), Thermo Scientific Chemicals
Catalog number013796.36
Price (USD)/ Each
989.00
-
Add to cart
Chemical Identifiers
CAS7440-47-3
IUPAC Namechromium
Molecular FormulaCr
InChI KeyVYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES[Cr]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
Appearance (Color)Gray
Comment-200 Mesh
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only