Thermo Scientific Chemicals

Sodium hydrogen sulfate monohydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 013799.A1
1 kg, Each
Thermo Scientific Chemicals

Sodium hydrogen sulfate monohydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 013799.A1
1 kg, Each
Quantity
Catalog number: 013799.A1
also known as 013799-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10034-88-5
IUPAC Name
sodium hydrate hydrogen sulfate
Molecular Formula
H3NaO5S
InChI Key
JXHZRQHZVYDRGX-UHFFFAOYSA-M
SMILES
O.[Na+].OS([O-])(=O)=O
Appearance (Color)
Colorless to white
Form
Crystals, moist in appearance
Free acid (titration)
Acidity (as H₂SO₄) 35.0-36.5%
Impurity content
As:≤1 ppm
Impurity content
Cl: ≤20 ppm

Description

Sodium hydrogen sulfate monohydrate is used in metal finishing and cleaning products. It is used to lower the pH of water for an effective chlorination in swimming pools. It is also used as a urine acidifier to reduce urinary stones in cats. Further, it is used in controlling outbreaks of crown-of-thorns starfish. It is a primary ingredient used in jewelry making to clean the surfaces. In addition to this, it is used as a food additive and serves as a reagent for the decomposition of basic metal oxides by fusion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text