Lithium, plasma standard solution, Specpure™ Li 1000μg/mL, Thermo Scientific Chemicals
Lithium, plasma standard solution, Specpure™ Li 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium, plasma standard solution, Specpure™ Li 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013821.AD
also known as 013821-AD
Price (USD)/ Each
45.65
Online exclusive
50.10 
Save 4.45 (9%)
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
45.65
Online exclusive
50.10 
Save 4.45 (9%)
Add to cart
Lithium, plasma standard solution, Specpure™ Li 1000μg/mL, Thermo Scientific Chemicals
Catalog number013821.AD
Price (USD)/ Each
45.65
Online exclusive
50.10 
Save 4.45 (9%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialLithium
CAS554-13-2
Health Hazard 1H290-H314-H335
Quantity50 mL
CAS7697-37-2
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only