Bismuth, plasma standard solution, Specpure™ Bi 1000μg/mL, Thermo Scientific Chemicals
Bismuth, plasma standard solution, Specpure™ Bi 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth, plasma standard solution, Specpure™ Bi 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
50 mL
500 mL
Catalog number 013822.AE
also known as 013822-AE
Price (USD)/ Each
63.65
Online exclusive
70.60 
Save 6.95 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
63.65
Online exclusive
70.60 
Save 6.95 (10%)
Add to cart
Bismuth, plasma standard solution, Specpure™ Bi 1000μg/mL, Thermo Scientific Chemicals
Catalog number013822.AE
Price (USD)/ Each
63.65
Online exclusive
70.60 
Save 6.95 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialBismuth
CAS7440-69-9
Health Hazard 1H290-H314-H335
Quantity100 mL
CAS7732-18-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only