Thermo Scientific Chemicals
Manganese, plasma standard solution, Specpure™ Mn 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 013826.AD
50 mL, Each
Thermo Scientific Chemicals
Manganese, plasma standard solution, Specpure™ Mn 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 013826.AD
50 mL, Each
Quantity
Catalog number: 013826.AD
also known as 013826-AD
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000?g/mL ?1%
Starting Material
Mn
Description
Manganese, plasma standard solution, Specpure™, Mn 1000μg/ml is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.