Thermo Scientific Chemicals

Platinum, plasma standard solution, Specpure™ 1000μg/mL, Thermo Scientific Chemicals

Catalog number: 013827.AD
50 mL, Each
Thermo Scientific Chemicals

Platinum, plasma standard solution, Specpure™ 1000μg/mL, Thermo Scientific Chemicals

Catalog number: 013827.AD
50 mL, Each
Quantity
Catalog number: 013827.AD
also known as 013827-AD
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000?g/mL ?1%
Starting Material
Platinum
Concentration
1,000 ? 3 ?g/ml Platinum
Matrix
20% HCl
Starting Material
Pt

Description

Platinum, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text