Platinum, plasma standard solution, Specpure™ 1000μg/mL, Thermo Scientific Chemicals
Platinum, plasma standard solution, Specpure™ 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum, plasma standard solution, Specpure™ 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
50 mL
500 mL
Catalog number 013827.AE
also known as 013827-AE
Price (USD)/ Each
136.65
Online exclusive
152.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
136.65
Online exclusive
152.00 
Save 15.35 (10%)
Add to cart
Platinum, plasma standard solution, Specpure™ 1000μg/mL, Thermo Scientific Chemicals
Catalog number013827.AE
Price (USD)/ Each
136.65
Online exclusive
152.00 
Save 15.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialPlatinum
CAS7440-06-4
Health Hazard 1H290-H314-H335
Quantity100 mL
CAS7647-01-0
View more
Platinum, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only