Palladium, plasma standard solution, Specpure™ Pd 1000μg/mL, Thermo Scientific Chemicals
Palladium, plasma standard solution, Specpure™ Pd 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium, plasma standard solution, Specpure™ Pd 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
50 mL
100 mL
Catalog number 013833.AP
also known as 013833-AP
Price (USD)/ Each
296.65
Online exclusive
329.00 
Save 32.35 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
296.65
Online exclusive
329.00 
Save 32.35 (10%)
Add to cart
Palladium, plasma standard solution, Specpure™ Pd 1000μg/mL, Thermo Scientific Chemicals
Catalog number013833.AP
Price (USD)/ Each
296.65
Online exclusive
329.00 
Save 32.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialPalladium, plasma standard solution, Specpure™
ConcentrationPd 1000μg/ml
Health Hazard 1H290-H314-H335
Quantity500 mL
Health Hazard 2GHS H Statement
H314-H335;
Causes severe skin burns and eye damage.
May cause respiratory irritation.
View more
Palladium, plasma standard solution finds application as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only