Thermo Scientific Chemicals
Dysprosium, plasma standard solution, Specpure™ Dy 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 013838.AE
100 mL, Each
Thermo Scientific Chemicals
Dysprosium, plasma standard solution, Specpure™ Dy 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 013838.AE
100 mL, Each
Quantity
Catalog number: 013838.AE
also known as 013838-AE
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000?g/mL ?1%
Starting Material
Dy2O3