Thermo Scientific Chemicals
Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 013839.AE
100 mL, Each
Thermo Scientific Chemicals
Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 013839.AE
100 mL, Each
Quantity
Catalog number: 013839.AE
also known as 013839-AE
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000?g/mL ?1%
Starting Material
Ni
Description
Nickel, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov