Thermo Scientific Chemicals

Beryllium plasma standard solution, Thermo Scientific Chemicals, Specpure™

Catalog number: 013848.AD
50 mL, Each
Thermo Scientific Chemicals

Beryllium plasma standard solution, Thermo Scientific Chemicals, Specpure™

Catalog number: 013848.AD
50 mL, Each
Quantity
Catalog number: 013848.AD
also known as 013848-AD
Price (USD)
Have Questions?
Starting Material
Be4O(C2H3O2)6
Concentration
1,000?g/mL ?1%
Concentration
1000?g/ml Beryllium
Matrix
5% HNO3

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text