Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL, Thermo Scientific Chemicals
Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
50 mL
100 mL
Catalog number 013852.AP
also known as 013852-AP
Price (USD)/ Each
167.00
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
167.00
Add to cart
Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL, Thermo Scientific Chemicals
Catalog number013852.AP
Price (USD)/ Each
167.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialCalcium
CAS471-34-1
Health Hazard 1H290-H314-H332-H335
Quantity500 mL
CAS7732-18-5
View more
Calcium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only