Lead, plasma standard solution, Specpure™ Pb 1000μg/mL, Thermo Scientific Chemicals
Lead, plasma standard solution, Specpure™ Pb 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead, plasma standard solution, Specpure™ Pb 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 013853.AD
also known as 013853-AD
Price (USD)/ Each
46.65
Online exclusive
51.80 
Save 5.15 (10%)
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
46.65
Online exclusive
51.80 
Save 5.15 (10%)
Add to cart
Lead, plasma standard solution, Specpure™ Pb 1000μg/mL, Thermo Scientific Chemicals
Catalog number013853.AD
Price (USD)/ Each
46.65
Online exclusive
51.80 
Save 5.15 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialLead
CAS7439-92-1
Health Hazard 1H290-H314-H335-H350-H360-H373
Quantity50 mL
CAS7697-37-2
View more
Lead, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only