Thermo Scientific Chemicals

Magnesium, plasma standard solution, Specpure™ Mg 1000μg/mL, Thermo Scientific Chemicals

Catalog number: 013861.AE
100 mL, Each
Thermo Scientific Chemicals

Magnesium, plasma standard solution, Specpure™ Mg 1000μg/mL, Thermo Scientific Chemicals

Catalog number: 013861.AE
100 mL, Each
Quantity
Catalog number: 013861.AE
also known as 013861-AE
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000?g/mL ?1%
Concentration
1000 ? 3?g/ml Magnesium
Matrix
5% HNO3
Starting Material
Magnesium
Starting Material
Mg

Description

Magnesium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text