Tin, plasma standard solution, Specpure™ Sn 1000μg/mL, Thermo Scientific Chemicals
Tin, plasma standard solution, Specpure™ Sn 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin, plasma standard solution, Specpure™ Sn 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013863.AD
also known as 013863-AD
Price (USD)/ Each
50.65
Online exclusive
55.70 
Save 5.05 (9%)
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
50.65
Online exclusive
55.70 
Save 5.05 (9%)
Add to cart
Tin, plasma standard solution, Specpure™ Sn 1000μg/mL, Thermo Scientific Chemicals
Catalog number013863.AD
Price (USD)/ Each
50.65
Online exclusive
55.70 
Save 5.05 (9%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialTin
Name NotePlasma standard solution, Sn 1000μg/mL
CAS7440-31-5
Health Hazard 1H290-H314-H335
Quantity50 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only