Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals
Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
50 mL
100 mL
Catalog number 013864.AP
also known as 013864-AP
Price (USD)/ Each
144.65
Online exclusive
161.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
144.65
Online exclusive
161.00 
Save 16.35 (10%)
Add to cart
Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals
Catalog number013864.AP
Price (USD)/ Each
144.65
Online exclusive
161.00 
Save 16.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialChromium
CAS7440-47-3
Health Hazard 1H314-H332-H335
Quantity500 mL
CAS7647-01-0
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only