Mercury, plasma standard solution, Specpure™ Hg 1000μg/mL, Thermo Scientific Chemicals
Mercury, plasma standard solution, Specpure™ Hg 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercury, plasma standard solution, Specpure™ Hg 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
50 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 013865.AE
also known as 013865-AE
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.50 
Save 10.85 (15%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.50 
Save 10.85 (15%)
Add to cart
Mercury, plasma standard solution, Specpure™ Hg 1000μg/mL, Thermo Scientific Chemicals
Catalog number013865.AE
Price (USD)/ Each
61.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
72.50 
Save 10.85 (15%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialMercury
CAS7439-97-6
Health Hazard 1H290-H314-H335-H373
Quantity100 mL
CAS7697-37-2
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only