Gallium, plasma standard solution, Specpure™ Ga 1000μg/mL, Thermo Scientific Chemicals
Gallium, plasma standard solution, Specpure™ Ga 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gallium, plasma standard solution, Specpure™ Ga 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013869.AD
also known as 013869-AD
Price (USD)/ Each
41.65
Online exclusive
46.60 
Save 4.95 (11%)
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
41.65
Online exclusive
46.60 
Save 4.95 (11%)
Add to cart
Gallium, plasma standard solution, Specpure™ Ga 1000μg/mL, Thermo Scientific Chemicals
Catalog number013869.AD
Price (USD)/ Each
41.65
Online exclusive
46.60 
Save 4.95 (11%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialGallium
CAS7440-55-3
Health Hazard 1H290-H314-H335
Quantity50 mL
CAS7647-01-0
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only