Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL, Thermo Scientific Chemicals
Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
50 mL
500 mL
Catalog number 013870.AE
also known as 013870-AE
Price (USD)/ Each
60.65
Online exclusive
67.50 
Save 6.85 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
60.65
Online exclusive
67.50 
Save 6.85 (10%)
Add to cart
Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL, Thermo Scientific Chemicals
Catalog number013870.AE
Price (USD)/ Each
60.65
Online exclusive
67.50 
Save 6.85 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialLanthanum
CAS1312-81-8
Health Hazard 1H290-H314-H335
Quantity100 mL
CAS7697-37-2
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only