Thermo Scientific Chemicals

Gold, plasma standard solution, Specpure™ Au 1000μg/mL, Thermo Scientific Chemicals

Catalog number: 013881.AP
500 mL, Each
Thermo Scientific Chemicals

Gold, plasma standard solution, Specpure™ Au 1000μg/mL, Thermo Scientific Chemicals

Catalog number: 013881.AP
500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 013881.AP
also known as 013881-AP
Price (USD)
Quantity
-
Have Questions?
Starting Material
Au
Starting Material
Gold
Concentration
1,000?g/mL ?1%
Matrix
20% HCl
Concentration
1,000 ?g/ml Gold

Description

Gold, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text