Thermo Scientific Chemicals

Dichlorobis(triphenylphosphine)nickel(II), 98%, Thermo Scientific Chemicals

Catalog number: 013930.09
10 g, Each
Thermo Scientific Chemicals

Dichlorobis(triphenylphosphine)nickel(II), 98%, Thermo Scientific Chemicals

Catalog number: 013930.09
10 g, Each
Quantity
Catalog number: 013930.09
also known as 013930-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14264-16-5
IUPAC Name
dichloronickelbis(ylium); bis(triphenylphosphane)
Molecular Formula
C36H30Cl2NiP2
InChI Key
VHQPRJDILRYZBG-UHFFFAOYSA-L
SMILES
Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Assay (unspecified)
> 97.0%
Appearance (Color)
Green to black
Form
crystalline powder

Description

Dichlorobis(triphenylphosphine)nickel(II) is used as a catalyst for cross-coupling of Grignard reagents, hydrosilylations, hydrogenation and polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Catalyst for cross-coupling of Grignard reagents, Hydrosilylations and hydrogenation, polymerizatron
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text