Thermo Scientific Chemicals

Silver trifluoroacetate, 98%, Thermo Scientific Chemicals

Catalog number: 013933.14
25 g, Each
Thermo Scientific Chemicals

Silver trifluoroacetate, 98%, Thermo Scientific Chemicals

Catalog number: 013933.14
25 g, Each
Quantity
Catalog number: 013933.14
also known as 013933-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2966-50-9
IUPAC Name
silver(1+) trifluoroacetate
Molecular Formula
C2AgF3O2
InChI Key
KZJPVUDYAMEDRM-UHFFFAOYSA-M
SMILES
[Ag+].[O-]C(=O)C(F)(F)F
Assay (unspecified)
97.6% min

Description

Silver trifluoroacetate is used as a Lewis acid catalyst. It is used as an important raw material for the preparation of triethylsilyl trifloroacetate and triphenylmethyl trifluoroacetate. Further, it is used as a catalyst in the preparation of 4-iodoveratrole from veratrole by reacting with iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text