Thermo Scientific Chemicals

Indium(III) phosphide, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog number: 013974.03
1 g, Each
Thermo Scientific Chemicals

Indium(III) phosphide, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog number: 013974.03
1 g, Each
Quantity
Catalog number: 013974.03
also known as 013974-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22398-80-7
IUPAC Name
indiganylidynephosphane
Molecular Formula
InP
InChI Key
GPXJNWSHGFTCBW-UHFFFAOYSA-N
SMILES
P#[In]
Total Metal Impurities
0.0001% max.

Description

Indium(III) phosphide is used in high-power and high-frequency electronics because of its superior electron velocity. It also has a direct band gap, making it useful for optoelectronics devices like laser diodes. It is also used as a substrate for epitaxial indium gallium arsenide based opto-electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text