Thermo Scientific Chemicals

Platinum(IV) sulfide, Pt 74.8% min, Thermo Scientific Chemicals

Catalog number: 013992.06
5 g, Each
Thermo Scientific Chemicals

Platinum(IV) sulfide, Pt 74.8% min, Thermo Scientific Chemicals

Catalog number: 013992.06
5 g, Each
Quantity
Catalog number: 013992.06
also known as 013992-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12038-21-0
IUPAC Name
disulfanylideneplatinum
Molecular Formula
PtS2
InChI Key
RKNFJIIYAUSTJA-UHFFFAOYSA-N
SMILES
S=[Pt]=S
Assay (unspecified)
Pt 74.8% min

Description

Ultra high purity and high purity compositions of platinum sulfide improve both optical quality and usefulness as scientific standards.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text