Titanium rod, 3.175mm (0.125in) dia, 99.99% (metals basis), Thermo Scientific Chemicals
Titanium rod, 3.175mm (0.125in) dia, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium rod, 3.175mm (0.125in) dia, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
5 cm
4 x 25 cm
Catalog number 014000.BU
also known as 014000-BU
Price (USD)/ Each
101.00
-
Add to cart
Quantity:
25 cm
Request bulk or custom format
Price (USD)/ Each
101.00
Add to cart
Titanium rod, 3.175mm (0.125in) dia, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number014000.BU
Price (USD)/ Each
101.00
-
Add to cart
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.99% (metals basis)
FormRod, 3.175mm (0.125in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only