Titanium wire, 2.0mm (0.08in) dia, 99.99% (metals basis), Thermo Scientific Chemicals
Titanium wire, 2.0mm (0.08in) dia, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium wire, 2.0mm (0.08in) dia, 99.99% (metals basis), Thermo Scientific Chemicals

Form: Approx. 14.2g/m; Temper: as drawn
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
Catalog number 014004.BU
also known as 014004-BU
Price (USD)/ Each
116.00
-
Add to cart
Quantity:
25 cm
Request bulk or custom format
Price (USD)/ Each
116.00
Add to cart
Titanium wire, 2.0mm (0.08in) dia, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number014004.BU
Price (USD)/ Each
116.00
-
Add to cart
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only