Barium peroxide, anhydrous, 84% min, Thermo Scientific Chemicals
Barium peroxide, anhydrous, 84% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium peroxide, anhydrous, 84% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
5 kg
Catalog number 014008.A1
also known as 014008-A1
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
Add to cart
Barium peroxide, anhydrous, 84% min, Thermo Scientific Chemicals
Catalog number014008.A1
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1304-29-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)84% min
Barium peroxide is a specialty product for proteomics research. It is a strong oxidizing agent. It is also used for bleaching in a variety of commercial applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only