Thermo Scientific Chemicals

Germanium(IV) iodide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 014009.18
50 g, Each
Thermo Scientific Chemicals

Germanium(IV) iodide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 014009.18
50 g, Each
Quantity
Catalog number: 014009.18
also known as 014009-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13450-95-8
Molecular Formula
GeH8I4
InChI Key
WTPFKACBVKJUIS-UHFFFAOYSA-N
SMILES
[GeH4].I.I.I.I
Molecular Weight (g/mol)
588.31
Total Metal Impurities
0.001% max.

Description

Germanium(IV) iodide is a biochemical for proteomics research. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text