Thermo Scientific Chemicals

Germanium(II) iodide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 014013.06
5 g, Each
Thermo Scientific Chemicals

Germanium(II) iodide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 014013.06
5 g, Each
Quantity
Catalog number: 014013.06
also known as 014013-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13573-08-5
Molecular Formula
GeH6I2
InChI Key
PUZROLYCAWSHNL-UHFFFAOYSA-N
SMILES
[GeH4].I.I
Molecular Weight (g/mol)
332.49
Total Metal Impurities
0.01% max.

Description

Germanium(II) iodide acts as a reagent that forms stable ionic adducts with carbenes. It is also used in the preparation of allylgermanes by reacting with allylstannanes. Further, it reacts with allyl bromide in the presence of zinc iodide to get allylgermanium(IV), which on reacts with carbonyl compounds to give the corresponding homoallylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text