Thermo Scientific Chemicals

Iron(II) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014024.09
10 g, Each
Thermo Scientific Chemicals

Iron(II) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014024.09
10 g, Each
Quantity
Catalog number: 014024.09
also known as 014024-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1317-37-9
IUPAC Name
λ²-iron(2+) sulfanediide
Molecular Formula
FeS
InChI Key
GNVXPFBEZCSHQZ-UHFFFAOYSA-N
SMILES
[S--].[Fe++]
Total Metal Impurities
0.1% max.

Description

Iron(II) sulfide is used to generate hydrogen sulfide instantaneously by reacting with hydrochloric acid. In laboratory, Kipp’s apparatus is utilized for this purpose. It is used as a re-sulphurizing and alloying agent, as a reducing agent to remove heavy metal impurities from phosphoric acid. It is used to control hydrogen embrittlement in alloy and stainless steel industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text