Thermo Scientific Chemicals

Silicon(IV) bromide, Thermo Scientific Chemicals

Catalog number: 014029.09
10 g, Each
Thermo Scientific Chemicals

Silicon(IV) bromide, Thermo Scientific Chemicals

Catalog number: 014029.09
10 g, Each
Quantity
Catalog number: 014029.09
also known as 014029-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-66-4
IUPAC Name
tetrabromosilane
Molecular Formula
Br4Si
InChI Key
AIFMYMZGQVTROK-UHFFFAOYSA-N
SMILES
Br[Si](Br)(Br)Br
Total Metal Impurities
0.1% max.

Description

Silicon(IV) bromide is used to grow thin films of silicon by the chemical vapor deposition method. It is also used for ceramics, sealants and the production of many cutting tools.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text