Thermo Scientific Chemicals

Potassium tungsten oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 014031.22
100 g, Each
Thermo Scientific Chemicals

Potassium tungsten oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 014031.22
100 g, Each
Quantity
Catalog number: 014031.22
also known as 014031-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-60-5
IUPAC Name
dipotassium dioxotungstenbis(olate)
Molecular Formula
K2O4W
InChI Key
AAQNGTNRWPXMPB-UHFFFAOYSA-N
SMILES
[K+].[K+].[O-][W]([O-])(=O)=O
Total Metal Impurities
0.5% max.

Description

Potassium tungsten oxide is used in solar energy and water treatment applications. It is used as alkaloids precipitating agent, bronze production and in tungsten refining. It is also used as an analytical reagent. Further, it is added to molten carbonate fuel cell to prevent the corrosion of stainless-steel separator and retards the sintering of the anode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text