Thermo Scientific Chemicals

Molybdenum(III) chloride, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 014034.04
2 g, Each
Thermo Scientific Chemicals

Molybdenum(III) chloride, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 014034.04
2 g, Each
Quantity
Catalog number: 014034.04
also known as 014034-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13478-18-7
IUPAC Name
molybdenum(3+) trichloride
Molecular Formula
Cl3Mo
InChI Key
ZSSVQAGPXAAOPV-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Mo+3]
Total Metal Impurities
0.5% max.

Description

Molybdenum(III) chloride reacts with tetrahydrothiophene to form complexes, which catalyze the polymerization and cyclotrimerization of alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text