Molybdenum(III) chloride, 99.5% (metals basis), Thermo Scientific Chemicals
Molybdenum(III) chloride, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum(III) chloride, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number 014034.09
Price (USD)/ Each
216.65
Online exclusive
240.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
216.65
Online exclusive
240.00 
Save 23.35 (10%)
Add to cart
Molybdenum(III) chloride, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number014034.09
Price (USD)/ Each
216.65
Online exclusive
240.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13478-18-7
IUPAC Namemolybdenum(3+) trichloride
Molecular FormulaCl3Mo
InChI KeyZSSVQAGPXAAOPV-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Mo+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Molybdenum(III) chloride reacts with tetrahydrothiophene to form complexes, which catalyze the polymerization and cyclotrimerization of alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only