Thermo Scientific Chemicals

Aluminum carbide, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 014038.18
50 g, Each
Thermo Scientific Chemicals

Aluminum carbide, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 014038.18
50 g, Each
Quantity
Catalog number: 014038.18
also known as 014038-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1299-86-1
IUPAC Name
1,3,5,7-tetraaluminahepta-3,4-dien-1,6-diyne
Molecular Formula
C3Al4
InChI Key
CAVCGVPGBKGDTG-UHFFFAOYSA-N
SMILES
[Al]#C[Al]=C=[Al]C#[Al]
Total Metal Impurities
0.9% max.

Description

Aluminum carbide is used as an abrasive material for cutting tools. It is also used to reduce metal oxides and to prepare aluminum nitride and methane. It and also finds use in metallurgy. Further, it acts as a catalyst and a drying agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text