Vanadium(III) chloride, 99% (metals basis), Thermo Scientific Chemicals
Vanadium(III) chloride, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium(III) chloride, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 014039.09
also known as 014039-09
Price (USD)/ Each
92.65
Online exclusive
103.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
92.65
Online exclusive
103.00 
Save 10.35 (10%)
Add to cart
Vanadium(III) chloride, 99% (metals basis), Thermo Scientific Chemicals
Catalog number014039.09
Price (USD)/ Each
92.65
Online exclusive
103.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7718-98-1
IUPAC Namevanadium(3+) trichloride
Molecular FormulaCl3V
InChI KeyHQYCOEXWFMFWLR-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[V+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Vanadium(III) chloride acts as an intermediate in organic synthesis and in the manufacture of organic light emitting diodes. It acts as a precursor to other vanadium(III) complexes. Further, it is used in the preparation of vanadium dichloride and organovanadium compounds. In addition to this, it serves as a catalyst in the production of ethylene propylene diene monomer rubber and polyethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only