Tantalum(V) fluoride, 99.9% (metals basis), Thermo Scientific Chemicals
Tantalum(V) fluoride, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantalum(V) fluoride, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 014052.09
also known as 014052-09
Price (USD)/ Each
144.65
Online exclusive
161.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
144.65
Online exclusive
161.00 
Save 16.35 (10%)
Add to cart
Tantalum(V) fluoride, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number014052.09
Price (USD)/ Each
144.65
Online exclusive
161.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7783-71-3
IUPAC Nametantalum(5+) pentafluoride
Molecular FormulaF5Ta
InChI KeyYRGLXIVYESZPLQ-UHFFFAOYSA-I
SMILES[F-].[F-].[F-].[F-].[F-].[Ta+5]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Tantalum(V) fluoride is used as a Friedel-Crafts catalyst. It combines with hydrogen fluoride and acts as a catalyst for the alkylation of alkanes and alkenes as well as the protonation of aromatic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only