Thermo Scientific Chemicals

Tantalum(V) fluoride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014052.18
50 g, Each
Thermo Scientific Chemicals

Tantalum(V) fluoride, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 014052.18
50 g, Each
Quantity
Catalog number: 014052.18
also known as 014052-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-71-3
IUPAC Name
tantalum(5+) pentafluoride
Molecular Formula
F5Ta
InChI Key
YRGLXIVYESZPLQ-UHFFFAOYSA-I
SMILES
[F-].[F-].[F-].[F-].[F-].[Ta+5]
Total Metal Impurities
0.1% max.

Description

Tantalum(V) fluoride is used as a Friedel-Crafts catalyst. It combines with hydrogen fluoride and acts as a catalyst for the alkylation of alkanes and alkenes as well as the protonation of aromatic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text