Barium fluoride, Optical Grade, Thermo Scientific Chemicals
Barium fluoride, Optical Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium fluoride, Optical Grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 014057.22
also known as 014057-22
Price (USD)/ Each
312.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
312.00
Add to cart
Barium fluoride, Optical Grade, Thermo Scientific Chemicals
Catalog number014057.22
Price (USD)/ Each
312.00
-
Add to cart
Chemical Identifiers
CAS7787-32-8
IUPAC Namebarium(2+) difluoride
Molecular FormulaBaF2
InChI KeyOYLGJCQECKOTOL-UHFFFAOYSA-L
SMILES[F-].[F-].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless or white
FormCrystals or powder or crystalline powder
Impurity content≤0.1% (≤1,000ppm)
Barium fluoride is used in ceramic flux, carbon brushes for electrical equipment, glass making, in the manufacture of other fluorides, crystals for spectroscopy, electronics and dry-film lubricants. Used for the antiseptic. For industry application, such as electron, measuring applicance, metallurgy, ceramics. Barium fluoride is often suitable for applications in the passive IR band (8 to 14 ?m) and is often used as a viewport window for thermography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only