Zirconium(IV) 2,4-pentanedionate, Thermo Scientific Chemicals
Zirconium(IV) 2,4-pentanedionate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium(IV) 2,4-pentanedionate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014065.30
Price (USD)/ Each
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS17501-44-9
IUPAC Namezirconium(4+) tetrakis(2,4-dioxopentan-3-ide)
Molecular FormulaC20H28O8Zr
InChI KeyDFPGFWYWSAYLCW-UHFFFAOYSA-N
SMILES[Zr+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
Insoluble material0.30% max ( in Toluene)
Assay (unspecified)0.30% max , percent volatiles
Formcrystalline solid
Melting Point188 to 195 deg C
Zirconium(IV) 2,4-pentanedionate is used as a cross-linking agent for oxygen-containing polymers. It is used as a catalyst in organic synthesis for the nitration of o-nitrotoluene using a dinitrogen pentoxide to get 2,4-dinitrotoluenes and 2,6-dinitrotoluenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only