Thermo Scientific Chemicals

Trichlorosilane, 98%, Thermo Scientific Chemicals

Catalog number: 014078.14
25 g, Each
Thermo Scientific Chemicals

Trichlorosilane, 98%, Thermo Scientific Chemicals

Catalog number: 014078.14
25 g, Each
Quantity
Catalog number: 014078.14
also known as 014078-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10025-78-2
IUPAC Name
trichlorosilyl
Molecular Formula
Cl3Si
InChI Key
PPDADIYYMSXQJK-UHFFFAOYSA-N
SMILES
Cl[Si](Cl)Cl
Assay (GC)
≥97.5%
Form
Liquid
Appearance (Color)
Clear colorless

Description

Trichlorosilane is used to prepare silicon for semiconductors. It is an active ingredient used in the preparation of polysilicons and silicone resins. It acts as coupling agents and silane adhesion promoter. It is a precursor to prepare organosilicon compounds through hydrosilylation reactions. Further, it is involved in the preparation of octadecyltrichlorosilane, perfluoroctyltrichlorosilane, and perfluordecyltrichlorosilane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text