Tetraethoxysilane, 99+%, Thermo Scientific Chemicals
Tetraethoxysilane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetraethoxysilane, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
Catalog number 014082.18
also known as 014082-18
Price (USD)/ Each
40.65
Online exclusive
45.30 
Save 4.65 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
40.65
Online exclusive
45.30 
Save 4.65 (10%)
Add to cart
Tetraethoxysilane, 99+%, Thermo Scientific Chemicals
Catalog number014082.18
Price (USD)/ Each
40.65
Online exclusive
45.30 
Save 4.65 (10%)
-
Add to cart
Chemical Identifiers
CAS78-10-4
IUPAC Nametetraethyl silicate
Molecular FormulaC8H20O4Si
InChI KeyBOTDANWDWHJENH-UHFFFAOYSA-N
SMILESCCO[Si](OCC)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
FormLiquid
Color (APHA)≤30
Assay from Supplier's CofA≥99.0%
Infrared spectrumConforms
As a binding agent and as a polymerization catalystTetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as a cross linking agent in silicon polymers and in aerogel preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only