Thermo Scientific Chemicals

Sodium metasilicate pentahydrate, tech., Thermo Scientific Chemicals

Catalog number: 014103.D9
5 x 1 kg, Each
Thermo Scientific Chemicals

Sodium metasilicate pentahydrate, tech., Thermo Scientific Chemicals

Catalog number: 014103.D9
5 x 1 kg, Each
Quantity
Catalog number: 014103.D9
also known as 014103-D9
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10213-79-3
IUPAC Name
disodium oxosilanebis(olate) pentahydrate
Molecular Formula
H10Na2O8Si
InChI Key
ADPGKKZKGXANON-UHFFFAOYSA-N
SMILES
O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O
Appearance (Color)
White
Form
Granular, -18 mesh
Assay (unspecified)
Silicon dioxide, SiO₂: 27.00-29.00%
Assay from Suppliers CofA
Sodium oxide, Na₂O: 28.00-30.50%
Particle Size
<1% retained on a +16 screen

Description

Sodium metasilicate pentahydrate is used as a starting material in the preparation of silicas and zeolites. It finds application in detergents and cleaners, pulp and paper production. It is also used in soil stabilization, sealing, adhesives and binders. Further, it is used in titanium dioxide production, paints for masonry and glass, spray coatings for tunnel construction. In addition to this, it is used in fruit and vegetable washes and sanitizers for food-contact surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text