Thermo Scientific Chemicals

Tin(II) oxalate, Thermo Scientific Chemicals

Catalog number: 014113.22
100 g, Each
Thermo Scientific Chemicals

Tin(II) oxalate, Thermo Scientific Chemicals

Catalog number: 014113.22
100 g, Each
Quantity
Catalog number: 014113.22
also known as 014113-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
814-94-8
Appearance (Color)
White
Form
Powder
Assay from Supplier's CofA
Tin (Sn): ≥55.7%

Description

Tin(II) oxalate is used for dyeing and printing textiles. It acts as a catalyst for the esterification reactions between long chain fatty acids and alcohols and in polymerization reactions. It is also used in stannous oral care compositions. Further, it is used in proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text